Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081056.1 | NP_001074525.1 | 18 - 19 | 2548 | 95 | |
GenBank mRNA | AK014235.1 | - | 1 - 2 | 114 | 95 | |
AK029113.1 | - | 13 - 14 | 2000 | 95 | ||
BC094337.1 | - | 3 - 4 | 567 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |