Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081037.2 | NP_001074506.1 | 9 - 10 | 1443 | 72 | |
NM_001242411.1 | NP_001229340.1 | 9 - 10 | 1443 | 72 | ||
GenBank mRNA | AY057898.1 | - | 2 - 3 | 274 | 72 | |
BC120892.1 | - | 9 - 10 | 1436 | 40061454 | 72 | |
BC120893.1 | - | 9 - 10 | 1436 | 40061455 | 72 | |
CX569081.1 | - | 1 - 2 | 202 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |