Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK050642.1 | - | 3 - 4 | 331 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |