Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163024.1 | NP_001156496.1 | 24 - 25 | 3449 | 106 | |
NM_001163025.1 | NP_001156497.1 | 23 - 24 | 3449 | 106 | ||
NM_153395.2 | NP_700444.2 | 23 - 24 | 3446 | 106 | ||
GenBank mRNA | AB014059.1 | - | 23 - 24 | 2968 | 106 | |
AK011826.1 | - | 2 - 3 | 237 | 106 | ||
AK012182.2 | - | 3 - 4 | 386 | 106 | ||
AK122425.1 | - | 24 - 25 | 3449 | 106 | ||
BC048406.1 | - | 11 - 12 | 1225 | 106 | ||
BC055354.1 | - | 6 - 7 | 692 | 106 | ||
BC150943.1 | - | 23 - 24 | 2970 | 9087854 | 106 | |
CV563363.1 | - | 2 - 3 | 309 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |