Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110015.1 | NP_001103485.1 | 6 - 7 | 768 | 65 | |
NM_001110016.1 | NP_001103486.1 | 6 - 7 | 768 | 65 | ||
NM_144863.4 | NP_659112.2 | 6 - 7 | 768 | 65 | ||
GenBank mRNA | AK041763.1 | - | 6 - 7 | 688 | 65 | |
AK045396.1 | - | 6 - 7 | 741 | 65 | ||
AK077545.1 | - | 6 - 7 | 698 | 65 | ||
AK140019.1 | - | 6 - 7 | 693 | 65 | ||
AK155965.1 | - | 6 - 7 | 667 | 65 | ||
AK156822.1 | - | 769 | 65 | |||
AK171683.1 | - | 6 - 7 | 706 | 65 | ||
AK172054.1 | - | 6 - 7 | 693 | 65 | ||
AK172377.1 | - | 6 - 7 | 698 | 65 | ||
BC022132.1 | - | 6 - 7 | 740 | 65 | ||
BC025809.1 | - | 6 - 7 | 682 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |