Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081403.1 | NP_001074872.1 | 1 - 2 | 956 | 86 | |
GenBank mRNA | AK018108.1 | - | 1 - 2 | 671 | 86 | |
AK173163.1 | - | 1 - 2 | 670 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |