Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026779.1 | NP_081055.1 | 13 - 14 | 2605 | 89 | |
GenBank mRNA | AK003797.1 | - | 4 - 5 | 410 | 89 | |
BC016223.1 | - | 13 - 14 | 2419 | 89 | ||
BC113180.1 | - | 13 - 14 | 2605 | 40060723 | 89 | |
BC113786.1 | - | 13 - 14 | 2605 | 40060721 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |