Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146084.1 | NP_666196.1 | 4 - 5 | 862 | 93 | |
GenBank mRNA | AK029766.1 | - | 4 - 5 | 854 | 93 | |
BC031465.1 | - | 4 - 5 | 862 | 3981146 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |