Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291804.1 | NP_001278733.1 | 14 - 15 | 2459 | 129 | |
NM_013504.5 | NP_038532.3 | 14 - 15 | 2459 | 129 | ||
GenBank mRNA | AK079441.1 | - | 14 - 15 | 2461 | 129 | |
AK081826.1 | - | 14 - 15 | 2367 | 129 | ||
BC109161.1 | - | 14 - 15 | 2296 | 40047883 | 129 | |
BC109162.1 | - | 14 - 15 | 2296 | 129 | ||
X97986.1 | - | 14 - 15 | 2393 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |