Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291804.1 | NP_001278733.1 | 3 - 4 | 569 | 90 | |
NM_013504.5 | NP_038532.3 | 3 - 4 | 569 | 90 | ||
GenBank mRNA | AK036893.1 | - | 3 - 4 | 455 | 90 | |
AK079441.1 | - | 3 - 4 | 570 | 90 | ||
AK081826.1 | - | 3 - 4 | 477 | 90 | ||
BC109161.1 | - | 3 - 4 | 406 | 40047883 | 90 | |
BC109162.1 | - | 3 - 4 | 406 | 90 | ||
X97986.1 | - | 3 - 4 | 503 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |