Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291809.1 | NP_001278738.1 | 11 - 12 | 1843 | 95 | |
NM_007882.3 | NP_031908.3 | 11 - 12 | 1843 | 95 | ||
GenBank mRNA | BC144845.1 | - | 11 - 12 | 1828 | 95 | |
CD773111.1 | - | 3 - 4 | 496 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |