Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291809.1 | NP_001278738.1 | 4 - 5 | 644 | 96 | |
NM_007882.3 | NP_031908.3 | 4 - 5 | 644 | 96 | ||
GenBank mRNA | BC144845.1 | - | 4 - 5 | 629 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |