Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026027.3 | NP_080303.1 | 1 - 2 | 36 | 61 | |
GenBank mRNA | AK002575.1 | - | 1 - 2 | 35 | 61 | |
AK003330.1 | - | 1 - 2 | 54 | 61 | ||
AK010532.1 | - | 1 - 2 | 50 | 61 | ||
AK012575.1 | - | 1 - 2 | 37 | 61 | ||
AK028148.1 | - | 1 - 2 | 42 | 61 | ||
BC024693.1 | - | 1 - 2 | 59 | 4020579 | 61 | |
BC094627.1 | - | 1 - 2 | 37 | 1532309 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |