Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013697.5 | NP_038725.1 | 1 - 2 | 259 | 65 | |
GenBank mRNA | AK018701.1 | - | 1 - 2 | 97 | 65 | |
AK050155.1 | - | 1 - 2 | 96 | 65 | ||
AK075588.1 | - | 1 - 2 | 96 | 65 | ||
BC024702.1 | - | 1 - 2 | 229 | 4192268 | 65 | |
BC032069.1 | - | 1 - 2 | 86 | 65 | ||
BC086926.1 | - | 1 - 2 | 96 | 6749152 | 65 | |
CK627038.1 | - | 1 - 2 | 85 | 65 | ||
D89076.1 | - | 1 - 2 | 96 | 65 | ||
X03351.1 | - | 1 - 2 | 96 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |