Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029491.2 | NP_083767.2 | 13 - 14 | 1964 | 145 | |
NM_177038.3 | NP_796012.2 | 13 - 14 | 1964 | 145 | ||
GenBank mRNA | AK041615.1 | - | 2 - 3 | 2051 | 145 | |
AK083747.1 | - | 13 - 14 | 1964 | 145 | ||
AK173085.1 | - | 6 - 7 | 741 | 145 | ||
BC048382.1 | - | 2 - 3 | 251 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |