Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081256.1 | NP_001074725.1 | 14 - 15 | 3841 | 85 | |
GenBank mRNA | AK129281.1 | - | 13 - 14 | 3197 | 85 | |
AK159090.1 | - | 1 - 2 | 174 | 85 | ||
BC038376.1 | - | 4 - 5 | 659 | 85 | ||
BC060727.1 | - | 7 - 8 | 1401 | 85 | ||
BC108415.1 | - | 7 - 8 | 1245 | 85 | ||
BC151084.1 | - | 14 - 15 | 3802 | 9087997 | 85 | |
BC172150.1 | - | 13 - 14 | 3202 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |