Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081256.1 | NP_001074725.1 | 7 - 8 | 1507 | 83 | |
GenBank mRNA | BC151084.1 | - | 7 - 8 | 1468 | 9087997 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |