Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144866.3 | NP_659115.3 | 3 - 4 | 449 | 103 | |
GenBank mRNA | AK049607.1 | - | 3 - 4 | 400 | 103 | |
AK077559.1 | - | 3 - 4 | 398 | 103 | ||
AK139933.1 | - | 3 - 4 | 410 | 103 | ||
AK151028.1 | - | 3 - 4 | 350 | 103 | ||
AK151710.1 | - | 3 - 4 | 350 | 103 | ||
AK152428.1 | - | 3 - 4 | 350 | 103 | ||
AK152952.1 | - | 3 - 4 | 350 | 103 | ||
AK164020.1 | - | 3 - 4 | 400 | 103 | ||
AK166740.1 | - | 3 - 4 | 418 | 103 | ||
AK169345.1 | - | 3 - 4 | 422 | 103 | ||
AK170221.1 | - | 3 - 4 | 392 | 103 | ||
BC013717.1 | - | 3 - 4 | 376 | 3992883 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |