Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204914.1 | NP_001191843.1 | 2 - 3 | 268 | 76 | |
NM_144865.2 | NP_659114.2 | 2 - 3 | 268 | 76 | ||
GenBank mRNA | AY562230.1 | - | 2 - 3 | 101 | 76 | |
BC020184.1 | - | 2 - 3 | 199 | 4505679 | 76 | |
CX216997.1 | - | 2 - 3 | 268 | 76 | ||
EU555417.1 | - | 2 - 3 | 101 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |