Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181414.5 | NP_852079.2 | 2 - 3 | 316 | 81 | |
GenBank mRNA | AK030578.1 | - | 2 - 3 | 344 | 81 | |
AK044997.1 | - | 2 - 3 | 316 | 81 | ||
BC027029.1 | - | 2 - 3 | 285 | 81 | ||
BC057678.1 | - | 2 - 3 | 319 | 3496633 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |