Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 26 - 27 | 3090 | 84 | |
GenBank mRNA | AK086942.1 | - | 25 - 26 | 2688 | 84 | |
AK136898.1 | - | 25 - 26 | 2689 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |