Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 27 - 28 | 3204 | 80 | |
GenBank mRNA | AK086942.1 | - | 26 - 27 | 2802 | 80 | |
AK136898.1 | - | 26 - 27 | 2803 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |