Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 6 - 7 | 869 | 60 | |
GenBank mRNA | AK086942.1 | - | 5 - 6 | 467 | 60 | |
AK132732.1 | - | 6 - 7 | 870 | 60 | ||
AK136898.1 | - | 5 - 6 | 468 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |