Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600014395.mm.1 |
TC0600001813.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014395.mm.1 |
NM_001177572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014395.mm.1 |
NM_001177572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014395.mm.1 |
NM_015829.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014395.mm.1 |
NM_015829.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014397.mm.1 |
TC0600001813.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014397.mm.1 |
NM_001177572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014397.mm.1 |
NM_001177572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014397.mm.1 |
NM_015829.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014397.mm.1 |
NM_015829.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014398.mm.1 |
TC0600001813.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014398.mm.1 |
NM_001177572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014398.mm.1 |
NM_001177572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014398.mm.1 |
NM_015829.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014398.mm.1 |
NM_015829.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014396.mm.1 |
TC0600001813.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014396.mm.1 |
NM_001177572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014396.mm.1 |
NM_001177572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014396.mm.1 |
NM_015829.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014396.mm.1 |
NM_015829.3 |
Refseq |