Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145398.2 | NP_663373.2 | 10 - 11 | 1464 | 64 | |
GenBank mRNA | AB091829.1 | - | 10 - 11 | 1464 | 64 | |
BC033520.1 | - | 8 - 9 | 1088 | 64 | ||
BC058953.1 | - | 2 - 3 | 457 | 64 | ||
BC125377.1 | - | 10 - 11 | 1387 | 40129892 | 64 | |
BC125379.1 | - | 10 - 11 | 1387 | 40129894 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |