Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF398972.1 | - | 1 - 2 | 234 | 95 | |
AK002412.1 | - | 1 - 2 | 207 | 95 | ||
AK005404.1 | - | 1 - 2 | 234 | 95 | ||
AK146664.1 | - | 1 - 2 | 234 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |