Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008748.3 | NP_032774.1 | 1 - 2 | 358 | 56 | |
GenBank mRNA | BC052705.1 | - | 1 - 2 | 116 | 5697787 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |