Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181315.4 | NP_851832.2 | 5 - 6 | 680 | 80 | |
GenBank mRNA | AK085670.1 | - | 5 - 6 | 655 | 80 | |
BC034413.1 | - | 5 - 6 | 631 | 4221296 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |