Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130513.1 | NP_001123985.1 | 14 - 15 | 2168 | 135 | |
NM_027286.4 | NP_081562.2 | 13 - 14 | 2020 | 135 | ||
GenBank mRNA | AB053181.1 | - | 13 - 14 | 2024 | 135 | |
AK008530.1 | - | 2 - 3 | 218 | 135 | ||
AK135357.1 | - | 8 - 9 | 1202 | 135 | ||
AK141604.1 | - | 13 - 14 | 1869 | 135 | ||
BC026801.1 | - | 13 - 14 | 2013 | 4236529 | 135 | |
EF408739.1 | - | 14 - 15 | 2129 | 135 | ||
EF408740.1 | - | 14 - 15 | 2129 | 135 | ||
GQ262785.1 | - | 13 - 14 | 1838 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |