Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029872.3 | NP_001025043.3 | 21 - 22 | 2650 | 79 | |
GenBank mRNA | AK156218.1 | - | 21 - 22 | 2651 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |