Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167891.1 | NP_001161363.1 | 1 | 372 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |