Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003671.1 | NP_001003671.1 | 1 | 329 | 74 | |
GenBank mRNA | AY013768.1 | - | 1 | 329 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |