Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010153.1 | NP_034283.1 | 3 - 4 | 421 | 77 | |
GenBank mRNA | AY686636.1 | - | 3 - 4 | 421 | 77 | |
BC106091.1 | - | 572 | 30844039 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |