Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019953.1 | NP_064337.1 | 4 - 5 | 748 | 80 | |
GenBank mRNA | AF186115.1 | - | 4 - 5 | 748 | 80 | |
AK007914.1 | - | 4 - 5 | 509 | 80 | ||
AK013014.1 | - | 4 - 5 | 619 | 80 | ||
AK013568.1 | - | 4 - 5 | 490 | 80 | ||
AK019927.1 | - | 4 - 5 | 620 | 80 | ||
AK150953.1 | - | 4 - 5 | 692 | 80 | ||
BC008261.1 | - | 4 - 5 | 520 | 2650612 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |