Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028873.4 | NP_083149.3 | 3 - 4 | 1598 | 72 | |
GenBank mRNA | AK017830.1 | - | 3 - 4 | 1612 | 72 | |
AK032681.1 | - | 3 - 4 | 1580 | 72 | ||
AK045345.1 | - | 3 - 4 | 1655 | 72 | ||
AK045445.1 | - | 3 - 4 | 1692 | 72 | ||
AK053314.1 | - | 3 - 4 | 1660 | 72 | ||
AK082579.1 | - | 3 - 4 | 1599 | 72 | ||
AK083878.1 | - | 3 - 4 | 1610 | 72 | ||
AK159388.1 | - | 3 - 4 | 1611 | 72 | ||
AK168935.1 | - | 3 - 4 | 1690 | 72 | ||
BC011146.1 | - | 3 - 4 | 1663 | 4016209 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |