Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010296.2 | NP_034426.2 | 6 - 7 | 705 | 121 | |
GenBank mRNA | BC131650.1 | - | 5 - 6 | 683 | 40110429 | 121 |
CV782971.1 | - | 1 - 2 | 99 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |