Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289617.1 | NP_001276546.1 | 1 - 2 | 289 | 103 | |
NM_001289618.1 | NP_001276547.1 | 1 - 2 | 289 | 103 | ||
NM_001289619.1 | NP_001276548.1 | 1 - 2 | 289 | 103 | ||
NM_011777.3 | NP_035907.1 | 1 - 2 | 289 | 103 | ||
GenBank mRNA | AK031415.1 | - | 1 - 2 | 88 | 103 | |
AK035743.1 | - | 1 - 2 | 60 | 103 | ||
AK076491.1 | - | 1 - 2 | 60 | 103 | ||
AK090110.1 | - | 1 - 2 | 56 | 103 | ||
AK147812.1 | - | 1 - 2 | 290 | 103 | ||
AK161709.1 | - | 1 - 2 | 85 | 103 | ||
AK170571.1 | - | 1 - 2 | 63 | 103 | ||
BC054775.1 | - | 1 - 2 | 248 | 6808149 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |