Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204273.1 | NP_001191202.1 | 4 - 5 | 373 | 93 | |
GenBank mRNA | AF204154.1 | - | 4 - 5 | 344 | 93 | |
AF204155.1 | - | 4 - 5 | 344 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |