Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00010525.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010525.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010525.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010528.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NR_033143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
NR_033143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010537.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010529.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010527.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NR_033143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
NR_033143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010538.mm.1 |
TC0X00001653.mm.1 |
MTA TC |