Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009793.3 | NP_033923.2 | 6 - 7 | 897 | 74 | |
GenBank mRNA | AK030219.1 | - | 6 - 7 | 781 | 74 | |
AK043114.1 | - | 6 - 7 | 607 | 74 | ||
BC070420.1 | - | 6 - 7 | 778 | 5718423 | 74 | |
X58995.1 | - | 6 - 7 | 587 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |