Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141737.1 | - | 7 - 8 | 803 | 73 | |
AK168287.1 | - | 7 - 8 | 798 | 73 | ||
BC004669.1 | - | 6 - 7 | 507 | 3498846 | 73 | |
U15562.1 | - | 7 - 8 | 769 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |