Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011766.5 | NP_035896.1 | 4 - 5 | 918 | 73 | |
GenBank mRNA | AF107306.1 | - | 4 - 5 | 439 | 73 | |
AF118845.1 | - | 4 - 5 | 688 | 73 | ||
AF125166.1 | - | 4 - 5 | 919 | 73 | ||
BC059241.1 | - | 4 - 5 | 818 | 6408975 | 73 | |
EF514219.1 | - | 4 - 5 | 1741 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |