Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019697.3 | NP_062671.1 | 3 - 4 | 1563 | 85 | |
GenBank mRNA | AB045326.1 | - | 2169 | 85 | ||
AB093280.1 | - | 3 - 4 | 2044 | 85 | ||
AF107780.1 | - | 3 - 4 | 1373 | 85 | ||
AK032268.1 | - | 2354 | 85 | |||
AK032772.1 | - | 2 - 3 | 139 | 85 | ||
BC079667.1 | - | 3 - 4 | 1563 | 30356567 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |