Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600014606.mm.1 |
TC0600001852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014606.mm.1 |
NM_001252266.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014606.mm.1 |
NM_001252266.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014606.mm.1 |
NM_010492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014606.mm.1 |
NM_010492.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014604.mm.1 |
TC0600001852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014604.mm.1 |
NM_001252266.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014604.mm.1 |
NM_001252266.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014604.mm.1 |
NM_010492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014604.mm.1 |
NM_010492.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014609.mm.1 |
TC0600001852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014609.mm.1 |
NM_001252266.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014609.mm.1 |
NM_001252266.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014609.mm.1 |
NM_010492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014609.mm.1 |
NM_010492.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014608.mm.1 |
TC0600001852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014608.mm.1 |
NM_001252266.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014608.mm.1 |
NM_001252266.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014608.mm.1 |
NM_010492.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014608.mm.1 |
NM_010492.3 |
Refseq |