Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009148.3 | NP_033174.2 | 1 - 2 | 85 | 84 | |
GenBank mRNA | AF022962.1 | - | 1 - 2 | 111 | 84 | |
AK010695.1 | - | 1 - 2 | 100 | 84 | ||
AK014499.1 | - | 1 - 2 | 108 | 84 | ||
AK036507.1 | - | 1 - 2 | 97 | 84 | ||
AK038125.1 | - | 1 - 2 | 108 | 84 | ||
AK039983.1 | - | 1 - 2 | 108 | 84 | ||
AK086603.1 | - | 1 - 2 | 104 | 84 | ||
AK173235.1 | - | 1 - 2 | 88 | 84 | ||
BC034644.1 | - | 1 - 2 | 85 | 5355276 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |