Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600003055.mm.1 |
TC0600003490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003055.mm.1 |
NM_178873.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003055.mm.1 |
NM_178873.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003058.mm.1 |
TC0600003490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003058.mm.1 |
NM_178873.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003058.mm.1 |
NM_178873.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003060.mm.1 |
TC0600003490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003060.mm.1 |
NM_178873.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003060.mm.1 |
NM_178873.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003057.mm.1 |
TC0600003490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003057.mm.1 |
NM_178873.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003057.mm.1 |
NM_178873.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003062.mm.1 |
TC0600003490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003062.mm.1 |
NM_178873.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003062.mm.1 |
NM_178873.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003059.mm.1 |
TC0600003490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003059.mm.1 |
NM_178873.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003059.mm.1 |
NM_178873.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003061.mm.1 |
TC0600003490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003061.mm.1 |
NM_178873.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003061.mm.1 |
NM_178873.3 |
Refseq |