Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700032024.mm.1 |
NM_001252530.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032024.mm.1 |
NM_001252530.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032024.mm.1 |
NM_001252531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032024.mm.1 |
NM_001252531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032024.mm.1 |
NM_175316.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032024.mm.1 |
NM_175316.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032024.mm.1 |
TC0700003804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032031.mm.1 |
NM_001252531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032031.mm.1 |
NM_001252531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032031.mm.1 |
TC0700003804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032029.mm.1 |
NM_001252531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032029.mm.1 |
NM_001252531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032029.mm.1 |
TC0700003804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032036.mm.1 |
NM_175316.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032036.mm.1 |
NM_175316.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032036.mm.1 |
TC0700003804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032023.mm.1 |
NM_001252530.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032023.mm.1 |
NM_001252530.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032023.mm.1 |
NM_001252531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032023.mm.1 |
NM_001252531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032023.mm.1 |
NM_175316.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032023.mm.1 |
NM_175316.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032023.mm.1 |
TC0700003804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032033.mm.1 |
NM_001252531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032033.mm.1 |
NM_001252531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032033.mm.1 |
TC0700003804.mm.1 |
MTA TC |