Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168382.1 | NP_001161854.1 | 3 - 4 | 1294 | 84 | |
NM_029404.3 | NP_083680.2 | 3 - 4 | 1291 | 84 | ||
GenBank mRNA | AK016517.1 | - | 3 - 4 | 1294 | 84 | |
BC040236.1 | - | 3 - 4 | 1257 | 3585881 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |