Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080285.1 | NP_525024.1 | 10 - 11 | 3234 | 64 | |
GenBank mRNA | AK032356.1 | - | 8 - 9 | 1615 | 64 | |
AK081137.1 | - | 6 - 7 | 1697 | 64 | ||
AK173254.1 | - | 10 - 11 | 3140 | 64 | ||
BC068156.1 | - | 10 - 11 | 2995 | 64 | ||
BC141407.1 | - | 10 - 11 | 3230 | 9056020 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |