Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177200.4 | NP_796174.2 | 13 - 14 | 1375 | 80 | |
GenBank mRNA | AK078436.1 | - | 3 - 4 | 345 | 80 | |
BC058741.1 | - | 13 - 14 | 1358 | 6309979 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |